Learn More
Corning™ LSE™ Adapters
Product Code. 15713338 Shop All Corning Products
Click to view available options
For Use With (Equipment):
Corning LSE Compact Centrifuge
Unit Size:
Pack of 6
Product Code. 15713338
Brand: Corning™ 480141
For use with Corning™ LSE™ Compact Centrifuge
This is an optional adapter for 5/7 mL tubes in 50 mL rotor, for use with Corning™ LSE™ Compact Centrifuge.
Order Info
dimForUseWithEquipment
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Adapter
For Use With (Equipment) Corning LSE Compact Centrifuge
For Use With (Application) Corning™ LSE™ Compact Centrifuge
No. per Pack 1 Each