Learn More
Eppendorf™ Adapters For Oak Ridge Tubes
Product Code. 10784872 Shop All Eppendorf Products
Click to view available options
Capacity:
10 mL
16 mL
30 mL
35 mL
Unit Size:
Pack of 2
Product Code. 10784872
Brand: Eppendorf™ 5820722002
Add flexibility to your liquid handling applications with these adapters
  • Adapters
  • For Eppendorf™ Oak Ridge Tubes
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Oak Ridge
Capacity 35 mL
For Use With (Equipment) 35mL Oak Ridge Tubes
No. per Pack 2 Pack