missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Diphosphate Disodium Salt, MP Biomedicals™

Product Code. p-7083072 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
This item is not returnable. View return policy

Product Code. 11388329

Brand: MP Biomedicals 02100056.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Involved in energy storage and nucleic acid metabolism. Adenosine-5'-Diphosphate Disodium Salt, MP Biomedicals is an adenine nucleotide that converts into ATP by ATP synthases. ADP affects platelet activation through its interaction with ADP receptors.

ADP affects platelet activation through its interaction with ADP receptors P2Y1, P2Y12 and P2X1. Upon its conversion to adenosine by ecto-ADPases, platelet activation is inhibited via adenosine receptors.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate
PubChem CID 85315
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Quantity 500 mg
Optical Rotation [α]20/D -29.6±1° c = 0.5% in 0.5M Na2HPO4 (Lit.)
Formula Weight 471.2
Percent Purity ≥95%
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.