missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adenylyl Cyclase Type V Inhibitor, NKY80
Click to view available options
Quantity:
5 mg
Description
A cell-permeable quinazolinone containing a non-nucleoside compound that acts as a potent, selective inhibitor of adenylyl cyclase (AC) type V isoform
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 5 mg |
Description | Pale orange |
Content And Storage | -30°C to -10°C |
For Use With (Application) | A cell-permeable quinazolinone containing a non-nucleoside compound that acts as a potent, selective inhibitor of adenylyl cyclase (AC) type V isoform |
CAS | 299442-43-6 |
Molecular Formula | C12H11N3O2 |
Formula Weight | 229.23 |
Synonym | NKY80; 2-Amino-7-(furan-2-yl)-7,8-dihydroquinazolin-5(6H)-one |
Sensitivity | Light Sensitive |
Solubility | Soluble in DMSO or methanol |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'