Learn More
Adipic acid, 99%, Thermo Scientific Chemicals
Catalog Number p-7021392
Click to view available options
Quantity:
1000 g
5000 g
25000 g
124-04-9
C6H10O4
146.142
MFCD00004420
WNLRTRBMVRJNCN-UHFFFAOYSA-N
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
196
CHEBI:30832
hexanedioic acid
C(CCC(=O)O)CC(=O)O
124-04-9
C6H10O4
146.142
MFCD00004420
WNLRTRBMVRJNCN-UHFFFAOYSA-N
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
196
CHEBI:30832
hexanedioic acid
C(CCC(=O)O)CC(=O)O
Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.
Solubility
Soluble in methanol and acetone. Slightly soluble in cyclohexane. Insoluble in benzene, acetic acid and petroleum ether.
Notes
Incompatible with oxidizing agents
TRUSTED_SUSTAINABILITY
pH 2.7
Melting Point 151°C to 155°C
Boiling Point ∼205°C (3mmHg)
Beilstein 1209788
Merck Index 14,162
Solubility Information Soluble in methanol and acetone. Slightly soluble in cyclohexane. Insoluble in benzene,acetic acid and petroleum ether.
Formula Weight 146.14
Percent Purity 99%
Density 1.36
Quantity 1000 g
Flash Point 196°C (384°F)
Odor Odorless
Chemical Name or Material Adipic acid