Learn More
Mecmesin™ Advanced Force Gauge
The Advanced Force Gauge (AFG) is the most versatile, fully-featured digital instrument.
1950.00€
Specifications
Height (Metric) 201 mm
Width (Metric) 70 mm
Weight (Metric) 650 g
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11977969
View Documents Promotion Details
Mecmesin™
850418
250 N
1950.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11997969
View Documents Promotion Details
Mecmesin™
850420
1000 N
1950.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
11937979
View Documents Promotion Details
Mecmesin™
850421
2500 N
1950.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
With ever-increasing demands for improved quality, durability and ease-of-use of all products, there is a growing need for more physical testing during the manufacturing processes. The Advanced Force Gauge (AFG) is a powerful instrument packed full of innovative features designed to suit a breadth of tension and compression tests.
The AFG offers enhanced test features and data analysis capabilities:
  • Max and 1st Peak mode - identify critical load points from the test
  • Fast data-acquisition speed - captures break-force readings accurately
  • Audible and visual pass/fail alarms - alerts operators immediately when testing
  • Overload warning with trend bar - to minimize accidental damage to loadcell
  • In-built storage for up to 500 readings - test rapidly now and download results later
  • Data transfer via RS232, digimatic or analog - output data to PC or printer for further analysis
  • Connection capability to external load cells and torque sensors - becomes a versatile display for many different applications
Specifications
Specifications
201 mm
70 mm
650 g