Learn More
Advanced Instruments Osmometer Standards
For in vitro diagonostic use
Brand:  Advanced Instruments 3MA040
Additional Details : Weight : 0.02000kg
Product Code. 15178616
  • 110.72€ / Pack of 10
Estimated Shipment: 05-06-2024
to see stock.
Description
Description
For in vitro diagonostic use
  • 2-Hydroxybiphenyl sodium salt tetrahydrate
  • 1% added as preservative
Specifications
Specifications
2°C to 30°C
2- Hydroxybiphenyl sodium salt tetrahydrate, 0.01% added as preservative
To check the performance of your osmometer and test methods, in-vitro diagnostic use
10 x 2 mL