missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AEE 788

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AEE 788 is a potent EGFR and VEGFR inhibitor (IC50 values are 2, 6, 59, 77, 160 and 330 nM for EGFR, ErbB2, VEGFR-1, VEGFR-2, ErbB4 and VEGFR-3 respectively). Also inhibits c-Abl, c-Fms and c-Src (IC50 values are 52, 60 and 61 nM respectively). Inhibits proliferation of EGFR and ErbB2 overexpressing cancer cell lines in vitro. Inhibits tumor xenograft growth and VEGF-induced angiogenesis in mice. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | AEE 788 |
| Purity | >98% |
| Molecular Weight (g/mol) | 440.58 |
| Inhibitors | AEE 788 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction