missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AF-DX 384

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent M2/M4 selective antagonist (pKi values are 8.22, 8.00, 7.51, 7.18 and 6.27 at human M2, M4, M1, M3 and M5 receptors respectively).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 478.64 |
| Purity | >98% |
| Molecular Weight (g/mol) | 478.641 |
| Chemical Name or Material | AF-DX 384 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction