Learn More
AF332 EP Color Scale
Used to assess product color by comparison with representative series of fixed color standards
Brand:  Lovibond Tintometer 433320
Additional Details : Weight : 2.47000kg
Product Code. 11914159
  • 2930.00€ / Each
Estimated Shipment: 05-08-2024
to see stock.
Récupel-bijdrage uitgesloten
Description
Description
  • Based on Lovibond Comparator System 2000, this kit is easy means of ordering complete range of equipment required for grading pharmaceutical solutions according to the EP Colour series
  • Includes five colour-grading discs
  • Each containing stable-coloured glass standards that match  reference solutions specified in European Pharmacopoeia
  • Sample (in 40mm glass cell) is viewed in Lovibond Comparator 2000+ with appropriate test disc until colour match is obtained
Specifications
Specifications
Lovibond™ Comparator System 2000, five color-grading discs and two 40 mm cells
Comparator 2000+
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products