Learn More
LNI Swissgas AG OFCAS iQ 200 10
Product Code. 17812159 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
17812159 100/115/220/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17812159 Supplier LNI Swissgas Supplier No. 6920.47.200
Regulatory: This product is under eco-contribution tax.
AG OFCAS-Scroll with dryer integrated.
  • 200 L/min at 10 bar, 230 V with dryer integrated.
Specifications
Voltage 100/115/220/230 V
Type Air Generator