missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Agar Bacteriological
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
- Agar is a solidifying agent used for culture media preparation, it is a purified agar from which the extraneous matter, pigmented portions and salts have been removed or reduced to a minimum.
Specifications
Specifications
| Content And Storage | The powder is very hygroscopic, store the powder at 10 to 30°C |
| Manufacturing Origin | Spain |
| Product Type | Agar |
| Sterility | Non-sterile |
| Fill Weight | 500g |
| Quantity | 500 g |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction