Learn More
Melford Agarose Low Melt
Description
Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.
Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.
Gel Strength, 1%: >200 g/cm2
Gelling Temperature: 26°C to 30°C
Melting Temperature: <65°C
Sulfate: <0.10%
EEO: <0.10
DNase and RNase: None Detected
Specifications
Specifications
| Chemical Name or Material | Agarose, Low Melt |
| CAS | 9012-36-6 |
| Quantity | 250 g |
| Grade | Molecular Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.