missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Agarose SS
Description
For DNA/RNA fragments >1000 bp.
For DNA/RNA fragments >1000 bp. Ideal for separating larger molecules. DNAse/RNase free.
Gel Strength, 1%: >1800 g/cm2
Gel Strength, 1.5%: >3200 g/cm2
Gelling Temperature: 35-37°C
Melting Temperature: 87-89°C
Sulfate: <0.12%
EEO: <0.13%
Specifications
Specifications
| Chemical Name or Material | Agarose SS |
| CAS | 9012-36-6 |
| Quantity | 100 g |
| Purity Grade Notes | Molecular Biology |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?