missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies AggreWell™ EB Formation Medium
Click to view available options
Quantity:
100 mL
Unit Size:
500mL
Description
- AggreWell™ EB Formation Medium is a serum-free medium that supports the survival of mTeSR™1-, TeSR™2-, or TeSR™-E8™-cultured human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells during generation and subsequent culture of embryoid bodies (EBs)
Specifications
Specifications
| Cell Type | Pluripotent Stem Cells |
| For Use With (Application) | Cell Differentiation |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction