Learn More
Trajan™ Split / Splitless Tapered FocusLiner for Agilent
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan™ 092011
Additional Details : Weight : 0.80000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent instruments
Specifications
GC Inlet Liner, Split/Splitless Tapered FocusLiner | |
78.5 mm | |
6.3 mm |
25/Pk. | |
4 mm | |
Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177 |