missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse PLUS High Voltage Output Cable

High Voltage Output Cable for use with AgilePulse PLUS.

Brand:  BTX™ 470095

Additional Details : Weight : 0.27000kg

Product Code. 16624862

  • 660.00€ / Each
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Cables and adapters are needed to connect an electrofusion/electroporation system to the electrodes and chambers to be used for a particular application.
  • BTX Electroporation systems are designed to be flexible when it comes to application, so users will sometimes need to buy additional connector cables when adding or changing sample accessory or electrodes.
Specifications

Specifications

AgilePulse PLUS
AgilePulse PLUS HV Output Cable, 5 ft.
152 cm
Chamber Cable
110/240 V
5 ft.
Non-sterile
Square Wave
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions