missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AH 7614
GREENER_CHOICE

Product Code. p-5545682
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18707076

Brand: Tocris Bioscience™ 5256/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective FFA4 (GPR120) antagonist

AH 7614 is a selective free fatty acid receptor 4 (FFA4/GPR120) antagonist (pIC50 values are 7.1 and <4.6 for human FFA4 and FFA1 receptors respectively). Inhibits linoleic acid and GSK 137647A -induced intracellular calcium accumulation in U2OS osteosarcoma cells expressing the FFA4 receptor.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type AH 7614
Purity >98%
Molecular Weight (g/mol) 351.42
Inhibitors AH 7614

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.