Learn More
Aalborg™ Air Mass Flowmeter
Product Code. 11822280
Click to view available options
Airflow Rate:
0 to 1 LPM
0 to 10 LPM
0 to 100 mL/min.
0 to 2 LPM
0 to 20 mL/min.
0 to 5 LPM
0 to 50 mL/min.
0 to 500 mL/min.
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11822280
Brand: Aalborg™ GFM17AVAL6A0/3NC06SS
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Type Mass Flowmeter
Accuracy ±5%
Airflow Rate 0 to 500 mL/min.
Certifications/Compliance NIST calibration certificate
Power Supply Details 12 V to 200 mA max
Precision ±1.5 %
Scale Full scale
Temperature (Metric) 5°C to 50°C