Learn More
Ismatec™ Air-Powered Double-Diaphragm PVDF Pump, Santoprene
Product Code. 15788398
Change view
Click to view available options
Flow Rate:
21 gal./min. max.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
15788398 21 gal./min. max. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15788398 Supplier Ismatec™ Supplier No. 7400021
Eliminates sparking concerns associated with electrical pumps.
  • Air-powered motor eliminates sparking concerns associate with electrical pumps.
  • Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing.
  • Non-metallic pumps are ideally suited for highly corrosive fluids intermittent/on-demand, portable low abrasive fluids, and suspended solids.
  • Safely handles abrasive fluids and slurries — ideal for high-viscosity and shear sensitive materials.
  • Encapsulated muffler provides excellent sound dampening.
Specifications
Description Air-powered, double-diaphragm
Flow Rate 21 gal./min. max.
Maximum Pressure 100 psi
Material Diaphragm: Santoprene PTFE
Temperature (Metric) Max 98°C
Temperature (Metric) Min. -12°C
Overall Length (Metric) 30 cm
Overall Width (Metric) 17.94 cm
Overall Height (Metric) 33.89 cm