Learn More
Al-23 Protective Tube, One End Closed, O.D.: 20mm, I.D.: 15mm, Wall.: 2.5mm, Thermo Scientific™
Product Code. 11372698
Click to view available options
Quantity:
200 mm
270 mm
375 mm
530 mm
740 mm
1030 mm
1430 mm
2030 mm
Unit Size:
1030mm
1430mm
200mm
2030mm
270mm
375mm
530mm
740mm
Product Code. 11372698
Brand: Thermo Scientific Alfa Aesar 030052.FA
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Protective Tube
Name Note One end closed
Quantity 2030 mm