Learn More
Al-23 Solid Rod, Round, Dia.: 4mm, Thermo Scientific™
Product Code. 11323078
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
250mm
300mm
600mm
800mm
Product Code. 11323078
Brand: Thermo Scientific Alfa Aesar 032929.DT
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Solid Rod
Quantity 250 mm
Form Rod