missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alexa Fluor™ 594 Tyramide SuperBoost™ Kit, goat anti-mouse IgG
Features poly-HRP-conjugated goat anti-mouse IgG secondary antibody. Alexa Fluor™ 488 Tyramide SuperBoost™ Kit combine brightness of AlexaFluor dyes with high signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce se
312.00€ - 610.00€
Specifications
Conjugate | Alexa Fluor 594 |
---|---|
Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
Product Line | SuperBoost™ |
Product Type | Tyramide Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15631892
|
Invitrogen™
B40915 |
Sufficient for 150 Slides |
610.00€
Pack of 150 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
15652232
|
Invitrogen™
B40942 |
Sufficient for 50 Slides |
312.00€
Pack of 50 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).Features:
- High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
- Simple protocol and detection using standard filters
- Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
- Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
Benefits:
- Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
- Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
- Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
- Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals
Shelf Life:
- Stable for 6 months after receiving
Specifications
Alexa Fluor 594 | |
Approved for shipment at Room Temperature or on Wet Ice | |
SuperBoost™ | |
Tyramide Kit |
For Research Use Only. Not for use in diagnostic procedures.