missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AlgiMatrix™ Dissolving Buffer
Click to view available options
Quantity:
50 mL
Unit Size:
Each
Description
AlgiMatrix™ Dissolving Buffer can be used with any of the AlgiMatrix™ 3D Culture Systems to dissolve sponges. It is safe to use with all cell types and will not disrupt cell spheroids, clusters or other 3D structures. Can be used right in the well or sponges can be moved to another vessel.
Specifications
Specifications
| Chemical Name or Material | Dissolving Buffer |
| For Use With (Equipment) | All Cell Types and Will Not Disrupt Cell Spheroids, Clusters or Other 3D Structures |
| Physical Form | Lyophilized |
| Product Line | Algimatrix |
| Quantity | 50 mL |
| Sterility | Sterile |
| Grade | Cell Culture Grade |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction