missing translation for 'onlineSavingsMsg'
Learn More

Alizarin-3-Methyliminodiacetic Acid, 85%, Honeywell Fluka™

Catalog Number p-7120135
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
3952-78-1
C19H15NO8
385.328
MFCD00001202
PWIGYBONXWGOQE-UHFFFAOYSA-N
alizarin complexone, alizarin fluorine blue, alizarin complexon, alizarine complexon, alizarine complexone, alizarinkomplexon, alizarine fluorine blue, 3-aminomethylalizarin-n,n-diacetic acid, az-c, alizarin-3-methyliminodiacetic acid
65132
CHEBI:53088
2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O
3952-78-1
C19H15NO8
385.328
MFCD00001202
PWIGYBONXWGOQE-UHFFFAOYSA-N
alizarin complexone, alizarin fluorine blue, alizarin complexon, alizarine complexon, alizarine complexone, alizarinkomplexon, alizarine fluorine blue, 3-aminomethylalizarin-n,n-diacetic acid, az-c, alizarin-3-methyliminodiacetic acid
65132
CHEBI:53088
2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O

for the determination of fluorine, ∼85%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 185°C
Color Yellow
Packaging Glass Bottle
Quantity 1 g
UN Number NONH for all modes of transport
Beilstein 2190028
Formula Weight 385.32g/mol
Percent Purity 85%
Chemical Name or Material Alizarin-3-Methyliminodiacetic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'