missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corticotropin Releasing Factor Antibody - BSA Free, Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
Corticotropin Releasing Factor Polyclonal specifically detects Corticotropin Releasing Factor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
Specifications
| Antigène | Corticotropin Releasing Factor |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Formule | PBS with 0.02% Sodium Azide |
| Alias de gène | corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor |
| Symboles de gène(s) | CRH |
| Espèces hôtes | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?