missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ALK [C1156Y], GST Tag, His Tag Recombinant Protein

Product Code. p-7121658
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15478823

Brand: Invitrogen™ PV6157

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Human ALK C1156Y, internal fragment, amino acids L1066-S1437 (as in NCBI/Protein entry NP_004295.2), C1156Y mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant

The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas. The translocation creates a fusion gene consisting of the ALK gene and the nucleophosmin gene: the 3' half of ALK, derived from chromosome 2, is fused to the 5' portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK. ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM HEPES with 100 mM NaCl, 15 mM glutathione, 20% glycerol, 5 mM DTT and no preservative; pH 7.5
Molecular Weight (g/mol) 70.5 kDa
Name Human ALK [C1156Y], GST Tag, His Tag
Quantity 10 μg
Common Name ALK (C1156Y Mutant)
Protein Length 1066-1437
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag, His-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.