Learn More
Alkali Metals, plasma standard solution, Specpure™, Ba,Be,Ca,Cs,K,Li,Mg,Na,Rb,Sr at 100μg/mL
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Alkali Metals, plasma standard solution |
| Name Note | Ba,Be,Ca,Cs,K,Li,Mg,Na,Rb,Sr at 100μg/mL |
| Physical Form | Liquid |
| MDL Number | MFCD08061357 |
| UN Number | UN3264 |
| Quantity | 100 mL |
| Grade | Specpure |