Learn More
Grace™ Alltech™ Vydac™ 201TP C18 Analytical Columns
Product Code. 11389550
Encompass
Click to view available options
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
Film Thickness:
5 μm
Unit Size:
Each
Product Code. 11389550
Brand: Grace™ 5103821
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Column Format: Analytical Column; Carbon Load: 8%; Pore Size: 300 Å; Column Phase: Reversed Phase
Specifications
Column Chemistry Aliphatic groups
Column Format Analytical Column
Diameter (Metric) 4.6 mm
Film Thickness 5 μm
Flow Rate 1 mL/min
For Use With LC/MS Separation
Length (Metric) 250 mm
Max. Pressure 1000 to 2000 psi (69 to 138 bar)
Mobile Phase Compatibility 15 : 75 : 10 Buffer/Methan/Methanol/n-Butanol; 2 : 88 : 10 Buffer/Methan/Methanol/n-Butanol; Buffer = Aqueous 0.02 M NH4OAc, pH 3.5
Particle Shape Spherical
Particle Size 5 μm
pH 3.5
Pore Size 300 Å
Surface Area 70 m²/g
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 8%
Column Type Reversed Phase
Endcapped No
Phase Reversed Phase
Type Analytical Column
Technique Reverse Phase HPLC