missing translation for 'onlineSavingsMsg'
Learn More

Allylpentafluorobenzene, 98%

Product Code. 11423428
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11423428

Brand: Thermo Scientific Alfa Aesar A17834.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 1736-60-3 | C9H5F5 | 208.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1736-60-3
Molecular Formula C9H5F5
Molecular Weight (g/mol) 208.13
MDL Number MFCD00000301
InChI Key YBDBTBVNQQBHGJ-UHFFFAOYSA-N
Synonym allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5
PubChem CID 74435
IUPAC Name 1,2,3,4,5-pentafluoro-6-prop-2-enylbenzene
SMILES FC1=C(F)C(F)=C(CC=C)C(F)=C1F

Specifications

Density 1.358
Boiling Point 148°C to 149°C
Flash Point 42°C (108°F)
Refractive Index 1.427
Quantity 5 g
UN Number UN1993
Beilstein 1971209
Formula Weight 208.13
Percent Purity 98%
Chemical Name or Material Allylpentafluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.