missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ALMS1 Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 7840 |
| Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-1195 |
| Gene Alias | Alstrom syndrome 1, Alstrom syndrome protein 1, DKFZp686A118, DKFZp686D1828, KIAA0328ALSS |
| Gene Symbol | ALMS1 |
| Quantity | 0.1 mg |
| Research Category | Immune System Diseases, Immunology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction