missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%

Product Code. p-7021688
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11468086

Brand: Thermo Scientific Alfa Aesar A14121.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13209-15-9 | C8H6Br4 | 421.752 g/mol

α,α,α',α'-Tetrabromo-o-xylene reacts with 5endo,6endo-bis-chlormethyl-norborn-2-en to obtain trans/cis-2,3-bis(chloromethyl)-1,4-methano-1,2,3,4-tetrahydroanthracene. Further, it is involved in the preparation of 1-(3-butenyl)-2-(deuteriomethyl)benzene by reacting with allylmagnesium bromide. In addition this, it is used to prepare exo-benzocyclobuteno-1,2,3,4-tetrahydro-1,4-methanoanthracene by reacting with exo-1,4,4a,8b-tetrahydro-1,4-methanobiphenylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α,α,α′,α′-Tetrabromo-o-xylene reacts with 5endo,6endo-bis-chlormethyl-norborn-2-en to obtain trans/cis-2,3-bis(chloromethyl)-1,4-methano-1,2,3,4-tetrahydroanthracene. Further, it is involved in the preparation of 1-(3-butenyl)-2-(deuteriomethyl)benzene by reacting with allylmagnesium bromide. In addition this, it is used to prepare exo-benzocyclobuteno-1,2,3,4-tetrahydro-1,4-methanoanthracene by reacting with exo-1,4,4a,8b-tetrahydro-1,4-methanobiphenylene.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13209-15-9
Molecular Formula C8H6Br4
Molecular Weight (g/mol) 421.752
MDL Number MFCD00000131
InChI Key LNAOKZKISWEZNY-UHFFFAOYSA-N
Synonym 1,2-bis dibromomethyl benzene, alpha,alpha,alpha',alpha'-tetrabromo-o-xylene, benzene, 1,2-bis dibromomethyl, unii-3wg2h68vx4, o-xylylene tetrabromide, alpha,alpha,alpha,alpha-tetrabromo-o-xylene, a,a,a',a'-tetrabromo-o-xylene, o-xylene, alpha,alpha,alpha',alpha'-tetrabromo, alpha,alpha,beta,beta-tetrabromo-o-xylene, acmc-209boi
PubChem CID 83234
IUPAC Name 1,2-bis(dibromomethyl)benzene
SMILES C1=CC=C(C(=C1)C(Br)Br)C(Br)Br

Specifications

Melting Point 114°C to 118°C
Quantity 50 g
UN Number UN1759
Beilstein 2211698
Solubility Information Insoluble in water.
Formula Weight 421.75
Percent Purity 97%
Chemical Name or Material alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.