missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha-Dichlorodiphenylmethane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
α,α-Dichlorodiphenylmethane is a degradation product of of polychloroaromatic insecticides such as Dicofol (D436700) and DDT. It is also used as a catalyst for photodegradation of plastics. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
α,α-Dichlorodiphenylmethane is a degradation product of of polychloroaromatic insecticides such as Dicofol (D436700) and DDT. It is also used as a catalyst for photodegradation of plastics. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Soluble in water (reacts), ether, benzene, ethanol, and toluene.
Notes
Stable under recommended storage conditions. Incompatible with strong oxidizing agents and bases.
α,α-Dichlorodiphenylmethane is a degradation product of of polychloroaromatic insecticides such as Dicofol (D436700) and DDT. It is also used as a catalyst for photodegradation of plastics. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Soluble in water (reacts), ether, benzene, ethanol, and toluene.
Notes
Stable under recommended storage conditions. Incompatible with strong oxidizing agents and bases.
Specifications
Density | 1.242 |
Boiling Point | 180°C to 181°C (17 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.605 |
Quantity | 100 g |
UN Number | UN3265 |
Solubility Information | Soluble in water (reacts),ether,benzene,ethanol,and toluene. |
Formula Weight | 237.13 |
Percent Purity | 97% |
Chemical Name or Material | alpha,alpha-Dichlorodiphenylmethane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'