missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications
Melting Point | ∼138°C |
Quantity | 5 g |
Solubility Information | Soluble in water. |
Formula Weight | 376.16 (304.10 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | alpha-D-Glucose-1-phosphate disodium salt tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'