missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog Number 11489952
136.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
56401-20-8
C6H13Na2O9P
306.114
MFCD00150839
AQMGMJDBUKKFOG-PKXGBZFFSA-N
alpha-d-glucose-1-phosphate disodium salt tetrahydrate
131855210
sodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
56401-20-8
C6H13Na2O9P
306.114
MFCD00150839
AQMGMJDBUKKFOG-PKXGBZFFSA-N
alpha-d-glucose-1-phosphate disodium salt tetrahydrate
131855210
sodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]

alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point ∼138°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 376.16 (304.10 Anhydrous)
Percent Purity 98%
Chemical Name or Material alpha-D-Glucose-1-phosphate disodium salt tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'