Learn More
Thermo Scientific™ alpha-Helical Corticotropin Releasing Factor (12-41)
182.00€ - 305.00€
Specifications
Content And Storage Store at -20°C
Format Solid
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15410547
SDS View Documents Promotion Details
Thermo Scientific™
J66287.!D
0.5 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15467589
SDS View Documents Promotion Details
Thermo Scientific™
J66287.M^
1 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Store at -20°C
Solid