Learn More
Ansell AlphaTec™ 1800 Ts PLUS
Product Code. 16824750
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
W18T111
W18T122
W18T156
Unit Size:
Pack of 30
Pack of 35
Pack of 40
Pack of 45
Product Code. 16824750
Brand: Ansell WH18T0015606
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Lightweight coveralls with breathable microporous laminate technology and Type 4/5/6 protection.
  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
  • Finger Loops
  • Chinstrap
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Seam Type Stitched, Taped
Certifications/Compliance (Europe) EN 374:2003, EN 14605:2005 + A1:2009,EN ISO 13982-1:2004 + A1:2010,EN 13034:2005 + A1:2009,EN 1073-2:2002,EN 1149-5:2008,EN 14126:2003
Includes Hood and Socks
For Use With (Application) Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders
Weight (Metric) 53 gsm
Style W18T156