Learn More
Ansell AlphaTec™ 2000 Lab Coats, Model 209
Product Code. p-7160713 Shop All Ansell Products
Click to view available options
Size:
Small
3X-Large
Unit Size:
Pack of 60
Product Code. 15915021
Brand: Ansell WH20B0020902
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Chemical resistant lab Coat features breathable microporous laminate technology.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Resistant Lab Coat
Size Small
Gender Unisex
Material Microporous Polyethylene Laminate
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
Style Standard Length
No. of Pockets 2
Certifications/Compliance CE Category III, EN 14126