missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Apron
Shop All Ansell Products
Click to view available options
Style:
W20B214
W20S213
Unit Size:
Pack of 200
Pack of 60
Size:
Small
Universal
X-Large
Description
- Silicone Free
- Low Linting
Specifications
Specifications
| For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support and Maintenance, Maintenance of Plant and Machines, Veterinary Services |
| Style | W20B214 |
| Product Type | Controlled Environments Gown |
| Material | Non-woven Microporous Polyethylene Laminate |
| Color | White |
| Closure Type | Tear-back |
| Disposable | Limited-use |
| Sterility | Non-sterile |
| Seam Type | Bound |
| Sleeve Style | Sleeveless |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction