missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Apron
Apron with protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards.
287.27€ - 474.55€
Specifications
| Product Type | Controlled Environments Gown |
|---|---|
| Material | Non-woven Microporous Polyethylene Laminate |
| Color | White |
| Sterility | Non-sterile |
| Product Code | Brand | Size | Style | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Size | Style | Price | Quantity & Availability | ||||
|
16130551
|
Ansell
WH20B-00214-02 |
Small | W20B214 |
684.00€ 474.55€ / Pack of 60 Save 209.45€ 30% Off |
In Stock
Log in to see stock.
|
||||
|
16110561
|
Ansell
WH20B-00214-05 |
X-Large | W20B214 |
722.00€ 474.55€ / Pack of 60 Save 247.45€ 34% Off |
Estimated Shipment: 17-12-2025
Log in to see stock. |
||||
|
16120561
|
Ansell
WH20B-00214-06 |
W20B214 |
684.00€ 474.55€ / Pack of 60 Save 209.45€ 30% Off |
Estimated Shipment: 27-01-2026
Log in to see stock. |
|||||
|
16130561
|
Ansell
WH20B-00214-07 |
W20B214 |
684.00€ 474.55€ / Pack of 60 Save 209.45€ 30% Off |
Estimated Shipment: 27-01-2026
Log in to see stock. |
|||||
|
16604423
|
Ansell
WH20S-00213-00 |
Universal | W20S213 |
415.00€ 287.27€ / Pack of 200 Save 127.73€ 30% Off |
Estimated Shipment: 27-01-2026
Log in to see stock. |
||||
Description
- Silicone Free
- Low Linting
Specifications
| Controlled Environments Gown | |
| Non-woven Microporous Polyethylene Laminate | |
| White | |
| Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title