Learn More
Ansell AlphaTec™ 2000 STANDARD Jacket & Trouser Set Bound
Product Code. p-7228400
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 40
Product Code. 17141015
Brand: Ansell WH20B0021903
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Superior protection from low hazard liquid spray and fine particulates with bound seams for improved overall protection.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Material Microporous Non-woven Polyethylene Laminate
Color White
Closure Type Zipper
Seam Type Bound
Certifications/Compliance (Europe) Partial Body Protection Only, EN 13034:2005 + A1:2009, DIN 32781:2010, EN 14126:2003, ASTM F1671, EN ISO 13688:2013
Product Type Chemical-resistant Jacket
Style W20B219
For Use With (Application) Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services