Learn More
Ansell AlphaTec™ 2500 STANDARD
Product Code. p-7228380
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
Pack of 25
Pack of 30
Product Code. 16844860
Brand: Ansell WH25W0012202
Only null left
Add to basket
Add to basket
Chemical protective suits with durable and breathable microporous laminate technology and Type 4/5/6 protection.
  • Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
  • Ultra low linting to help reduce the risk of cross contamination in critical areas
  • Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Color White
For Use With (Application) Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments
Hood Type Attached
Material Microporous Non-woven Polypropylene Laminate
Size Small
Product Type Ultrasonically Welded Coverall
Seam Type Welded
Style W25W122
Weight (Metric) 65 gsm