Learn More
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 103
Product Code. p-200007429 Shop All Ansell Products
Click to view available options
Size:
3X-Large
4X-Large
5X-Large
Unit Size:
Each
Product Code. 17999140
Brand: Ansell YE30W0010307
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
AlphaTec™ 3000 Ultrasonically Welded Model 103 features a durable, comfortable multi-layer chemical barrier with dual-zip systems and double cuffs.
  • Reinforced protection: The AlphaTec™ 3000 Ultrasonically Welded - Model 103 protective suit features multi-layered fabric that ensures an effective barrier against numerous inorganic chemicals and biohazards
  • Enhanced features: This protective barrier is further reinforced by its double cuffs, dual-zip systems and double zip enclosure, which extends wear life
  • Increased visibility: Thanks to its highly visible bright yellow color, this protective outer garment remains particularly noticeable in low-lit environments, enhancing wearer and workplace safety
  • Assured antistatic properties: In addition, it complies with EN 1149-5 antistatic standards
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size 3X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Double
Neck Style Collared
Hood Type Unhooded
Closure Type Double Zipper
Seam Type Welded
Sterility Non-sterile
Protection Class Type 3-B, Type 4-B, Type 5-B, Type 6-B
Resistance Liquid, Chemicals, Particle, Anti-static, Bloodborne Pathogens
Construction Coverall with Collar, Double Zip Closure, Double Cuffs, Elasticated Waist, Double Cuffs and Ankles
Certifications/Compliance (Europe) EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, CE Number, EN 1073-2:2002, EN 1149-5:2018, EN ISO 13688:2013, EN 14126:2003
Latex Free No
No. of Pockets None
For Use With (Application) Transferring liquids and solids between vessels and tanks and process equipment, Unexpected leakages spills or other releases, Opening and draining pumps valves or lines, Loading and unloading trucks and vehicles, Filling, blending and charging of raw materials, Product inspection, Mixing or loading chemicals, Cleaning of plant and machines
Style 3000 103