missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 4000 AVANT2 AIRline Ultrasonically Welded and Taped G02

Product Code. p-200004967
Click to view available options
Size:
2X-Large
3X-Large
Large
Large/X-Large
Medium
Small
Unit Size:
Pack of 3
This item is not returnable. View return policy

Product Code. 17160795

Brand: Ansell™ GR40T00075702G02

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

AlphaTec™ 4000 AVANT2 AIRline Ultrasonically welded and taped G02 is affixed by the wearer and detachable for reuse when appropriate. The air flow is adjustable from 340 L/min to 590 L/min at a working pressure of 3.5 to 5 bar.

Affixed by the wearer and detachable for reuse.

  • The air flow is adjustable from 340 L/min to 590 L/min at a working pressure of 3.5 to 5 bar.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Exhalation valves fitted to the rear of the suit allows C0to escape and equalises pressure within the suit, allowing for a full range of movement without risk of excessive pressure causing harm to the suit or the wearer
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 14605:2005 + A1:2009|CE CATEGORY III|EN 1073-1:2016 + A1:2018|EN 1149-5:2018
Color Green
For Use With (Application) Contaminated Environments|Chemical Handling|Filling, blending and charging of raw materials|Transferring liquids and solids|Opening and draining pumps valves or lines|Loading and unloading of proces equipment|Raw material sample collection|Concentration mixing and palletisation|Hazmat response|Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Small
Ankle Style Attached Socks with Elasticated Boot Overflap
Closure Type Zipper
Collar Type Encapsulated Hood
Cuff Style Elasticated double cuffs with finger and thumb loop and permanently attached chemical protective gloves
Disposable Reusable
For Use With (Equipment) Air-fed/Supplied Systems
Gender Unisex
Neck Style Fully Encapsulated Hood
No. of Pockets None
Product Type Encapsulated chemical protective suits for use with air-fed/supplied systems
Protection Class CE CATEGORY III
Resistance Chemicals I Particle I Antistatic
Seam Type Welded and Taped
Sterility Non-sterile
Waist Style Polyester webbing belt with YKK buckle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.