Learn More
Euro Protection™ Altaite Low-Cut Safety Shoes
Product Code. p-7146787
Click to view available options
Size:
37
40
41
42
43
44
45
47
Unit Size:
Each
Product Code. 12023676
Brand: Euro Protection™ 9ALTL37
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Euro Protection™ Altaite Low-Cut Safety Shoes are comfortable, snug, and durable, and feature a composite toe cap for protection.
  • Padded collar and bellows tongue for snug fit.
  • Heel scuff cap
  • Shock-absorbing EVA midsole.
  • Full grain leather and bright mesh combination upper.
  • Breathable 3D mesh lining.
  • Removable, textile and foam insole.
Specifications
Color Brown, Beige
Liner Material Breathable, 3D Mesh
Material Leather
Size 37
Sole Features Cemented, shock-absorbing EVA midsole, nitrile rubber outsole, non-metallic (composite fabric) puncture-resistant sole.
Sole Material Nitrile, EVA, Composite Fabric
Style Low
Toe Material Composite
Weight (Metric) 603 g per shoe in size 8
Certifications/Compliance (Europe) EN ISO 20345 : 2011