Learn More
Lovibond™ Aluminum #2 tablets
Click to view available options
Quantity:
100 Tablets
250 Tablets
Unit Size:
Each
Pack of 100
Description
ECR method for comparators and photometers
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
Specifications
| Quantity | 250 Tablets |
| Range | Aluminum #2 0.01 to 0.30 mg/L |
| Type | Reagent |
Safety and Handling
Product Identifier
- Aluminium No. 2
- Warning
- Flammable solid Category 2
- Skin sensitiser Category 1
- H228-Flammable solid.
- H317-May cause an allergic skin reaction.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : methenamine