Learn More
Aluminum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Catalog Number p-7073200
Click to view available options
Quantity:
100 mL
500 mL
134029
134029
Aluminum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M
TRUSTED_SUSTAINABILITY
Packaging HDPE plastic bottle
Quantity 100 mL
Color Colorless
pH 1
Physical Form Liquid
compliance-icons
Product Identifier
  • Aluminium solution 1000 ppm
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid, Aluminum nitrate