missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Mg, AA 5182, A95182, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Aluminum, Al/Mg, AA 5182, A95182, Ø65x25mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Beryllium/Antimony/Zirconium/Gallium/Bismuth/Lead/Nickel/Strontium/Vanadium/Tin/Titanium/Chromium/Zinc/Copper/Silicon/Iron/Manganese/Magnesium/Cobalt/Silver/Phosphorus/Sodium/Arsenic/Calcium/Boron/Aluminium |
| CAS | 7429-90-5 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0012 %/Beryllium 0.002 %/Antimony 0.0034 %/Zirconium 0.0051 %/Gallium 0.0065 %/Bismuth 0.0096 %/Lead 0.019 %/Nickel 0.024 %/Strontium 0.025 %/Vanadium 0.033 %/Tin 0.034 %/Titanium 0.042 %/Chromium 0.063 %/Zinc 0.096 %/Copper 0.105 %/Silicon 0.25 %/Iron 0.316 %/Manganese 0.39 %/Magnesium 4.01 %/Cobalt (<2) %/Silver (<3) %/Phosphorus (<30) %/Sodium (<30) %/Arsenic (10) %/Calcium (20) %/Boron (8) %/Aluminium (94.6) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction