Learn More
Aluminum carbide, 99+% (metals basis)
Description
Aluminum carbide is used as an abrasive material for cutting tools. It is also used to reduce metal oxides and to prepare aluminum nitride and methane. It and also finds use in metallurgy. Further, it acts as a catalyst and a drying agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2100°C (>2200° (decomposition)) |
| Color | Gray |
| Quantity | 250 g |
| UN Number | UN1394 |
| Merck Index | 14,335 |
| Solubility Information | Soluble in dilute hydrochloric acid. |
| Formula Weight | 143.96 |
| Percent Purity | ≥99% |
| Sensitivity | Moisture sensitive |
| Density | 2.36 g/mL |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.