Learn More
Bochem™ Aluminum Chemical Scoop
Product Code. 10554841
Click to view available options
Capacity (Metric):
10 mL
65 mL
90 mL
240 mL
400 mL
480 mL
900 mL
1.6 L
Length (Metric):
52 mm
145 mm
155 mm
250 mm
270 mm
285 mm
290 mm
310 mm
350 mm
Unit Size:
Each
Product Code. 10554841
Brand: Bochem™ 8827
Only null left
Add to basket
Add to basket
Manufactured from aluminum, the Bochem™ Aluminum Chemical Scoop – is a perfect addition to your laboratory tools.
  • Lightweight
  • Aluminum
  • Length is total for combined scoop and handle
Specifications
Capacity (Metric) 90 mL
Product Type Chemical Scoop
Material Aluminum
Length (Metric) 250 mm